Order Entry
Austria
Orders LinkContactUsLinkComponent
Human IL-10 ELISPOT Set

Human IL-10 ELISPOT Set

Human IL-10 ELISPOT Set
Catalog # ABCAAB46549-480
Supplier:   Abcam
Human IL-10 ELISPOT Set
Catalog # ABCAAB46549-480
Supplier:  Abcam
CAS Number:  
Human IL-10 ELISPOT Set
Human IL-10 ELISPOT Set

Human IL-10 ELISPOT Set

CAS Number:  
Price
1 110,00 €
Log in for availability
1 * 480 Tests
Product Details & Documents

Human IL-10 ELISPOT Set is a ELISPOT Set for the sensitive detection of Human IL-10 production in a single cell suspension.

Specifications

Specifications

  • Description:
    Human IL-10 ELISPOT Set
  • Cat. No.:
    ABCAAB46549-480
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Species reactivity:
    Human
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen symbol:
    IL-10

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.