Orders LinkContactUsLinkComponent
Permanent marker
Permanent marker
Permanent marker, fiber tip, Green
Catalog # KOEH2-5071
Permanent marker
Catalog # KOEH2-5071
Permanent marker
Permanent marker, fiber tip, Green
Price
18,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
This versatile paint marker dispense a fast-drying, high-gloss enamel paint for permanent marking, lettering, coding and numbering of both interior and exterior surfaces of metal, plastic, printed circuit boards, glass, wood, paper, leather, textiles, etc. This marker can write on porous or non-porous; rough or smooth; wet, oily or dry surfaces.
  • Marks are permanent
  • Withstand heat and weathering under adverse conditions
  • Available in different lead-free colours
  • With long-lasting replaceable fiber tips
Specifications
  • Colour:
    Green
  • Description:
    Permanent marker, fiber tip
  • Seq:
    04