Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Lead(II) acetate basic

Glass bottle for solids, 500 g
Catalog # A16763.36
Health
Environment
Lead(II) acetate basic
Catalog # A16763.36
CAS Number:  1335-32-6
Lead(II) acetate basic
Lead(II) acetate basic

Glass bottle for solids, 500 g

CAS Number:  1335-32-6
Price
83,30 €
Log in for availability
1 * 500 g
Product Details & Documents

CAS No.: 1335-32-6 Molecular Formula: C4H6O4Pb.2Pb(OH)2 Formula Weight: 807.72 Merck Reference: 14,5419 UN No.: 1616 MDL No.: MFCD00013041

Synonyms:  Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
  • CAS Number:1335-32-6
  • Formula:(CH₃COO)₂Pb·2Pb(OH)₂
  • MDL Number:MFCD00011154
  • Molecular Weight:807.72 g/mol
  • Merck Index:14,5419
  • Storage Temperature:Ambient
  • EINECS:215-630-3
  • UN:1616
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications