Order Entry
Austria
Orders LinkContactUsLinkComponent
Lead(II) acetate basic

Lead(II) acetate basic

Plastic bottle for solids, 2.500 g
Catalog # A16763.0E
Health
Environment
Lead(II) acetate basic
Catalog # A16763.0E
CAS Number:  1335-32-6
Lead(II) acetate basic
Lead(II) acetate basic

Plastic bottle for solids, 2.500 g

CAS Number:  1335-32-6
Price
325,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

CAS No.: 1335-32-6 Molecular Formula: C4H6O4Pb.2Pb(OH)2 Formula Weight: 807.72 Merck Reference: 14,5419 UN No.: 1616 MDL No.: MFCD00013041

Synonyms:  Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
  • CAS Number:1335-32-6
  • Formula:(CH₃COO)₂Pb·2Pb(OH)₂
  • MDL Number:MFCD00011154
  • Molecular Weight:807.72 g/mol
  • Merck Index:14,5419
  • Storage Temperature:Ambient
  • EINECS:215-630-3
  • UN:1616
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    2.500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.