Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Syringes for CTC analytics systems

Fixed needle, 22, 500 µl, 51 mm, 0,72 mm, LC, 500F-CTC-GT-LC (0.4) *
Catalog # 554-1464
Syringes for CTC analytics systems
Catalog # 554-1464
Syringes for CTC analytics systems
Syringes for CTC analytics systems

Fixed needle, 22, 500 µl, 51 mm, 0,72 mm, LC, 500F-CTC-GT-LC (0.4) *

Price
118,00 €
Log in for availability
1 * 1 ST
Specifications
  • Ext.Ø:
    0,72 mm
  • Gauge:
    22
  • Type:
    500F-CTC-GT-LC (0.4) *
  • Type of tip:
    LC
  • Volume:
    500 µl
  • Description:
    Fixed needle
  • Length:
    51 mm
  • Seq:
    09
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.