Order Entry
Austria
Orders LinkContactUsLinkComponent
Syringes for CTC analytics systems

Syringes for CTC analytics systems

Fixed needle, 26, 250 µl, 50 mm, 0,47 mm, Conical, 250F-CTC-GT-5/0.47C *
Catalog # 554-1461
Syringes for CTC analytics systems
Catalog # 554-1461
Syringes for CTC analytics systems
Syringes for CTC analytics systems

Fixed needle, 26, 250 µl, 50 mm, 0,47 mm, Conical, 250F-CTC-GT-5/0.47C *

Price
129,00 €
Log in for availability
1 * 1 ST
Specifications
  • Ext.Ø:
    0,47 mm
  • Gauge:
    26
  • Type:
    250F-CTC-GT-5/0.47C *
  • Type of tip:
    Conical
  • Volume:
    250 µl
  • Description:
    Fixed needle
  • Length:
    50 mm
  • Seq:
    46
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.