Dissection boards, DissecTable™
Small board with 3 cutting areas, White, 330,2×279,4×12,5 mm
Supplier: Simport Scientific
Jump to:
Product Details & Documents
HDPE / PS. These dissection boards feature a large cutting area on one side. They include dimensional scales in inches and centimeters, a grid and dimensional circles of different sizes. The other side offers two cutting boards half the size with the same dimensional features printed on each one of them.
- Drain groove is carved all around the edge of the boards in order to contain fluids
- All corners have rubber feet giving more stability to the working surface
- Bases are stackable and will not move sideways during dissecting work
Documents
Specifications
- Colour:White
- Description:Small board with 3 cutting areas
- W×D×H:330,2×279,4×12,5 mm