pH buffers, colour coded, reference standards, Supelco®
4,00, ±0,01, Red, 25 °C, Plastic bottle, 1×500 ml, 500 mL
Supplier: Merck
pH buffers, colour coded, reference standards, Supelco®
Catalog # B5020-500ML
Supplier: Merck
CAS Number:
Springe zu:
Product Details & Documents
Certifications: Standardised against N.I.S.T. standard reference materials
Documents
Spezifikationen
Spezifikationen
- Pack type:Plastic bottle
- Pk:500 mL
- Reference temperature:25 °C
- Colour:Red
- pH accuracy:±0,01
- Pk Info:1×500 ml
- pH value:4,00
- Cat. No.:B5020-500ML
- M-Clarity quality level:MQ100



