Order Entry
Austria
Orders LinkContactUsLinkComponent
 

pH buffers, colour coded, reference standards, Supelco®

10,00, ±0,01, Blue, 25 °C, Plastic bottle, 6×500 ml, 6
  B4895-6X500ML
 :   Merck
pH buffers, colour coded, reference standards, Supelco®
  B4895-6X500ML
 :  Merck
 :  
pH buffers, colour coded, reference standards, Supelco®
pH buffers, colour coded, reference standards, Supelco®

10,00, ±0,01, Blue, 25 °C, Plastic bottle, 6×500 ml, 6

 :  
 
235,00 €
   
1 * 6 ST
 

 : Standardised against N.I.S.T. standard reference materials

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    6
  • Reference temperature:
    25 °C
  • Colour:
    Blue
  • pH accuracy:
    ±0,01
  • Pk Info:
    6×500 ml
  • pH value:
    10,00
  • Cat. No.:
    B4895-6X500ML
  • M-Clarity quality level:
    MQ100

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.