Orders LinkContactUsLinkComponent
Microtubes
single, for use with racks, 0,65 ml
  RATI8200021
 :   RATIOLAB
Microtubes
  RATI8200021
 :  RATIOLAB
Microtubes
Microtubes
single, for use with racks, 0,65 ml
213,80 €
1 * 5.000 ST
Translucent polypropylene (PP) with very low protein binding.
  • Very good temperature resistance
  • Very good chemical resistance
The 0.65 and 1.2 ml micro-tubes can be supplied as single, strips of 8 or in 96-well format as a refill system for 8 x 12 plates.
  • Capacity:
    0,65 ml
  • Description:
    single, for use with racks