Order Entry
Austria
Orders LinkContactUsLinkComponent
Microtubes

Microtubes

single, for use with racks, 1,2 ml
Catalog # RATI8200001
Supplier:   RATIOLAB
Microtubes
Catalog # RATI8200001
Supplier:  RATIOLAB
Microtubes
Microtubes

single, for use with racks, 1,2 ml

Price
213,80 €
Log in for availability
1 * 5.000 ST
Product Details & Documents

Translucent polypropylene (PP) with very low protein binding.

  • Very good temperature resistance
  • Very good chemical resistance

The 0.65 and 1.2 ml micro-tubes can be supplied as single, strips of 8 or in 96-well format as a refill system for 8 x 12 plates.

Specifications
  • Capacity:
    1,2 ml
  • Description:
    single, for use with racks