Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing cassette, System II Hex

Tissue processing cassette, System II Hex

Pink
Catalog # 720-1895
Supplier:   CELLPATH
Tissue processing cassette, System II Hex
Catalog # 720-1895
Supplier:  CELLPATH
Tissue processing cassette, System II Hex
Tissue processing cassette, System II Hex

Pink

Price
67,90 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

The System II Hex Cassette differs from conventional round hole processing cassettes in that it has hexagonal instead of round perforations arranged in a honeycombed structure to better utilise the available surface area.

  • Available in different stock colours, which allow laboratories to colour code different types of workload or different applications
  • Manufactured from high–grade virgin acetyl resin

This unique arrangement increases the open area by approximately 13 percent thereby increasing reagent flow through and improving specimen staining and analysis time.

A potential cost saving benefit is that there is less carry over of chemicals so reducing the frequency of chemical change.

Although made to an international industry standard size, the Hex Cassette is designed to suit all established cassette–writing machines and benefits from extra thick side walls, so increasing stability and rigidity, which is crucial for high quality sectioning.

Specifications
  • Colour:
    Pink
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.