Orders LinkContactUsLinkComponent
Pycnometer
Pycnometer
100 ml
Catalog # HECH42570040
Pycnometer
Catalog # HECH42570040
Pycnometer
Pycnometer
100 ml
Price
41,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in stopper
  • Good chemical resistance
  • Good temperature resistance
Certifications: DIN ISO 3507
Specifications
  • Volume (nom.):
    100 ml