Rubber Stoppers
35,0 mm, 31,0 mm, 38,0 mm
HECH41590038
DIN 12871, made of natural rubber, hardness 40 Shore A.
- Ø bottom:31,0 mm
- Ø top:38,0 mm
- Height:35,0 mm
- Seq:09
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



