Dissecting scissors
sharp/sharp, 155 mm
Catalog # MART11-300-15
Supplier: KLS Martin Group
Jump to:
Product Details & Documents
Dissecting scissors made of stainless steel. One model features a TC Gold finish.
- TC Gold finish: Tungsten Carbide inlays provide an exceptionally long service life
- Straight
Specifications
- Type:sharp/sharp
- Length:155 mm