Orders LinkContactUsLinkComponent
Dissecting scissors
Curved, sharp/sharp, 115 mm
  HAMML-HSB301-11
Dissecting scissors
  HAMML-HSB301-11
Dissecting scissors
Dissecting scissors
Curved, sharp/sharp, 115 mm
318,00 €
1 * 1 ST
These dissecting scissors made of stainless steel feature tungsten carbide inserts.
  • Type:
    Curved, sharp/sharp
  • Length:
    115 mm
  • Seq:
    02