ContactUsLinkComponent
TEST SIEVE SHAKER EML 200 DIGITAL PLUS T FOR DRY-SIEVING 230 VOLT, 50 HERTZ INCLUDING CLAMPING SYSTEM CLASSIC AND A COVER WITH INSP ECTION GLASS 1 * 1 items
Catalog # 510-0948
Supplier:  NEXOPART
TEST SIEVE SHAKER EML 200 DIGITAL PLUS T FOR DRY-SIEVING 230 VOLT, 50 HERTZ INCLUDING CLAMPING SYSTEM CLASSIC AND A COVER WITH INSP ECTION GLASS 1 * 1 items
Catalog # 510-0948
Supplier:  NEXOPART
Frequently Bought Together