Filter sets
Filter set 38, excitation BP 470/40, beam splitter FT 495, emission BP 525/50
Catalog # ZEIS0000001031346
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Description:Filter set 38, excitation BP 470/40, beam splitter FT 495, emission BP 525/50
- Seq:08