Order Entry
Austria
Orders LinkContactUsLinkComponent
CellStor Pot™ specimen container

CellStor Pot™ specimen container

60 ml, 10% formal saline
Catalog # 720-2323
Supplier:   CELLPATH
CellStor Pot™ specimen container
Catalog # 720-2323
Supplier:  CELLPATH
CellStor Pot™ specimen container
CellStor Pot™ specimen container

60 ml, 10% formal saline

Price
Inquire For Price
Log in for availability
1 * 25 ST
Product Details & Documents

Prefilled specimen containers.

  • Leak-proof lid for safe transportation and specimen storage
  • With traceable badge number, safety data sheet in the packaging
Specifications
  • Capacity:
    60 ml
  • Description:
    10% formal saline
  • Seq:
    03