Orders LinkContactUsLinkComponent
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
CellSafe™ biopsy capsule
Catalog # 720-2334
Supplier:   CELLPATH
Biopsy specimen processing, CellSafe™
Catalog # 720-2334
Supplier:  CELLPATH
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
CellSafe™ biopsy capsule
Price
630,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
  • No tissue damage
  • Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
Specifications
  • Description:
    CellSafe™ biopsy capsule