Cooling racks
Cooling rack, PP, natural, for tubes up to 1,5 or 2,0 ml, 64, 130×130×57 mm
Catalog # GLWAG02
Supplier: GLW
Jump to:
Product Details & Documents
These cooling racks are made of polypropylene for test tubes that require continued cooling.
- Sturdy construction
- Autoclavable up to +121 °C
- Cover must be ordered seperately
Remove the insert, add chrushed ice, reinsert and start working. Use in freezer down to −90° C
Specifications
- No. of Holes:64
- Description:Cooling rack, PP, natural, for tubes up to 1,5 or 2,0 ml
- W×D×H:130×130×57 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cover for cooling racks, PP, natural, autoclavableCover for cooling racks, PP, natural, autoclavableGLWAGD1Log in for availability1 * 1 ST - 1,96 €
Product Information
- Catalog #AvailabilityPrice
- Cover for cooling racks, PP, black, autoclavable
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



