Box 10 x 10, yellow, 140 x 153 x 53 mm, compartment size 12.5 mm x 12.5 mm, made of polypropylene, autoclavable at 121c and use in freezer down to -90c 1 * 1 items
479-1528
: GLW
Box 10 x 10, yellow, 140 x 153 x 53 mm, compartment size 12.5 mm x 12.5 mm, made of polypropylene, autoclavable at 121c and use in freezer down to -90c 1 * 1 items
479-1528
: GLW
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK