Orders LinkContactUsLinkComponent
Certified colour reference standards
Certified colour reference standards
Pt-Co 0 mg Pt/l, certified under ISO 9001 quality system, Glass bottle, 500 mL
Catalog # TINT133991
Supplier:   Lovibond Tintometer
Certified colour reference standards
Catalog # TINT133991
CAS Number:  
Certified colour reference standards
Certified colour reference standards
Pt-Co 0 mg Pt/l, certified under ISO 9001 quality system, Glass bottle, 500 mL
CAS Number:  
Price
99,60 €
Log in for availability
1 * 500 mL
Product Details & Documents
For routine calibration of colorimeters and verification of test data.
Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.
Specifications
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Pt-Co 0 mg Pt/l, certified under ISO 9001 quality system