NITRATES STANDARD SOLUTION AT 10G/L DAN 1 * 100 mL
Catalog # CMPL6FEB.10K.W
Supplier: CUSTOM MADE CHEMICALS LAB
NITRATES STANDARD SOLUTION AT 10G/L DAN 1 * 100 mL
Catalog # CMPL6FEB.10K.W
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection