Order Entry
Austria
Orders LinkContactUsLinkComponent
D-(-)-Mannitol ≥96%, powder

D-(-)-Mannitol ≥96%, powder

1 kg
Artikelnr. ICNA0210224801
Anbieter:   MP Biomedicals
D-(-)-Mannitol ≥96%, powder
Artikelnr. ICNA0210224801
Anbieter:  MP Biomedicals
CAS Nummer:  69-65-8
D-(-)-Mannitol ≥96%, powder
D-(-)-Mannitol ≥96%, powder

1 kg

CAS Nummer:  69-65-8
Preis
126,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Mannitol is carbohydrate found in plants and plant exudates.

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Soluble in water (1 g/5,5 ml), alcohol (1 g/83 ml), glycerol (1 g/18 ml); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:163…172 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8

Documents

Specifications

Specifications

  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.