Order Entry
Austria
Orders LinkContactUsLinkComponent
Bath fluids

Bath fluids

SilOil, −90...+55 °C, >90 °C, 3 mm²/s, 5 L
Catalog # HUBE6258
Supplier:   HUBER
Bath fluids
Catalog # HUBE6258
Supplier:  HUBER
CAS Number:  
Bath fluids
Bath fluids

SilOil, −90...+55 °C, >90 °C, 3 mm²/s, 5 L

CAS Number:  
Price
915,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Thermal fluids with a wide range of operating temperatures and good thrmodynamic properties for use in circulators and heating systems where water is not appropriate.

All viscosities values are dynamic visosities measured at 25 °C except DW-Therm® which was measured at 20 °C.

Caution: * –20...+235 °C for closed system applications

Use biocides safely. Always read the label and product information before use

Specifications

Specifications

  • Pk:
    5 L
  • Flash point:
    >90 °C
  • Temperature range:
    −90...+55 °C
  • Type:
    SilOil
  • Viscosity:
    3 mm²/s

Specifications