Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR®, Zero Air Generators

VWR®, Zero Air Generators

7 l/min, 280×330×304 mm
Catalog # VWRU26000-024
Supplier:   VWR Collection
VWR®, Zero Air Generators
Catalog # VWRU26000-024
Supplier:  VWR Collection
VWR®, Zero Air Generators
VWR®, Zero Air Generators

7 l/min, 280×330×304 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Designed as a compact, hazard free, lower cost alternative to the user of expensive high pressure gas cylinders in the laboratory. Replacing cylinders with a gas generator brings advantages : increased safety, greater convenience, reduced cost and higher purity gas for more accurate analysis.

  • Produces zero air from compressed air
  • Ensures a flat and consistent base line
  • Short payback period

Purity: <0,05 ppm THC.

Inlet pressure 2,8 to 8,6 bar.

Inlet/outlet port size: ¹/₄" NPT female.

Specifications
  • Flow rate:
    7 l/min
  • Seq:
    01
  • W×D×H:
    280×330×304 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.