Orders LinkContactUsLinkComponent
Charts
Charts
C180, –30...+50 °C, 24 hours
Catalog # 620-1498
Supplier:   DICKSON
Charts
Catalog # 620-1498
Supplier:  DICKSON
Charts
C180, –30...+50 °C, 24 hours
Price
79,10 €
Log in for availability
1 * 60 ST
Product Details & Documents
Documents
Specifications
  • Description:
    C180
  • Seq:
    06
  • Range:
    –30...+50 °C
  • Recording time:
    24 hours