Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR® Surface Sampling Sponge Kits

VWR® Surface Sampling Sponge Kits

Dehydrated surface sampling sponges in a 450 ml bag
Catalog # 710-1020
Supplier:   VWR Chemicals
VWR® Surface Sampling Sponge Kits
Catalog # 710-1020
Supplier:  VWR Chemicals
VWR® Surface Sampling Sponge Kits
VWR® Surface Sampling Sponge Kits

Dehydrated surface sampling sponges in a 450 ml bag

Price
792,00 €
Log in for availability
1 * 400 ST
Product Details & Documents

Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sponge sterile sampling bag, with a foil-sealed, pre-moistened cellulose sponge, and 10 ml of sterile neutralising buffer.

  • Sterilised using gamma irradiation and leakproof transport bags, for the rapid cooling of samples in refrigerators
  • Puncture-proof safety bags eliminating sharp points that can cause puncture and/or abrasions
  • Larger surface in comparison with normal sampling swabs

This surface sampling sponge kit is designed to collect and transport samples, to detect the presence of microbiological contaminations such as Listeria, Salmonella, E. coli, and other food-borne pathogens on almost any surface. These kits are widely used in the food, medical, public health and cosmetic industries.

Certifications: HACCP, USDA, ACIA and CFIA requirements.

Caution: Gloves should be worn during all sampling procedures.

Specifications
  • Description:
    Dehydrated surface sampling sponges in a 450 ml bag
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.