Adapters, cone/screw thread, Quickfit®
24/29 NS, 24 mm, 10,5 - 11,5 mm
Catalog # 201-0227
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer. Ideal for securing thermometers, gas inlet tubes etc.
Specifications
- Cone size:24/29 NS
- Hole Ø:10,5 - 11,5 mm
- Thread:24 mm
- Seq:08



