Order Entry
Austria
Orders LinkContactUsLinkComponent
Containers, rectangular

Containers, rectangular

8,5 l, 570×350×70 mm, White, PP
Catalog # SEMA1294
Supplier:   SEMADENI
Containers, rectangular
Catalog # SEMA1294
Supplier:  SEMADENI
Containers, rectangular

8,5 l, 570×350×70 mm, White, PP

Price
36,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

HDPE or PP, white, stackable.

These are ideal containers with side handles and edge for gripping to handle solid samples. Containers over 225 mm height are not suitable for liquids.

Specifications
  • Capacity:
    8,5 l
  • Colour:
    White
  • Material:
    PP
  • Seq:
    02
  • W×D×H:
    570×350×70 mm