ContactUsLinkComponent
qScript cDNA SuperMix provides a sensitive and easy-to-use solution for two-step RT-PCR. This 5X concentrated master mix provides a ll necessary components (except RNA template) for first-strand synthesis including: buffer, dNTPs, MgCl2, primers, RN 1 * 5 items
Catalog # 733-1179
Supplier:  Quantabio
undefined
qScript cDNA SuperMix provides a sensitive and easy-to-use solution for two-step RT-PCR. This 5X concentrated master mix provides a ll necessary components (except RNA template) for first-strand synthesis including: buffer, dNTPs, MgCl2, primers, RN 1 * 5 items
Catalog # 733-1179
Supplier:  Quantabio
Frequently Bought Together