Order Entry
Austria
Orders LinkContactUsLinkComponent
Squeegees, Plastic

Squeegees, Plastic

Plastic squeegee with foam blade, Yellow, 610 mm
Catalog # MICRSQYE-24
Supplier:   MICRONOVA
Squeegees, Plastic
Catalog # MICRSQYE-24
Supplier:  MICRONOVA
Squeegees, Plastic
Squeegees, Plastic

Plastic squeegee with foam blade, Yellow, 610 mm

Price
131,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These plastic, double foam squeegees are easy to sanitise.

  • Will not rust
  • Extremely durable
  • Synthetic rubber handle with PP frame
  • For use with HACCP compliance programs and colour coding

Integrated blade eliminates seams and crevices, reducing places for bacteria to grow. Frame handle is threaded for use with STA series of plastic handles.

Specifications
  • Colour:
    Yellow
  • Description:
    Plastic squeegee with foam blade
  • Length:
    610 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.