Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Buffer solutions

Buffer standard DIN 19266, 1,679, -, 25 °C, Plastic bottle, 500 mL
Catalog # REGE101679
Supplier:   REAGECON
Buffer solutions
Catalog # REGE101679
Supplier:  REAGECON
CAS Number:  
Buffer solutions
Buffer solutions

Buffer standard DIN 19266, 1,679, -, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
41,70 €
Log in for availability
1 * 500 mL
Product Details & Documents

 

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Description:
    Buffer standard DIN 19266
  • pH accuracy:
    -
  • pH value:
    1,679

Specifications