L(+)-Ascorbic acid, Sigma-Aldrich®
Glass bottle for solids, 50 g
Catalog # SIAL95210-50G
Supplier: Merck
Springe zu:
Product Details & Documents
Synonyme: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Nummer:50-81-7
- Formel:C₆H₈O₆
- MDL-Nummer:MFCD00064328
- Molekulargewicht:176.13 g/mol
- Merck-Index:13,00837
- Siedepunkt:553 °C (1013 hPa)
- Schmelzpunkt:190…192 °C
- Dichte:1.694 g/cm³ (20 °C)
- Lagertemperatur:Ambient
- EINECS:200-066-2
- REACH:exempt
Spezifikationen
Spezifikationen
- Pack type:Glass bottle for solids
- Pk:50 g



