Order Entry
Austria
Orders LinkContactUsLinkComponent
Eukitt® mounting medium

Eukitt® mounting medium

Eukitt® mounting medium, Glass bottle, 500 mL
Catalog # 03989-500ML
Eukitt® mounting medium
Catalog # 03989-500ML
CAS Number:  
Eukitt® mounting medium

Eukitt® mounting medium, Glass bottle, 500 mL

CAS Number:  
Price
187,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

This is a mounting medium (contains 45% acrylic resin and 55% xylenes) used in microscopy. It is an adhesive and specimen preservative that can be used manually and in automated coverslipping equipment. Special purpose adhesive and sealant for use with glass, non plastic and other non-reactive materials.

  • Fast-drying (20 minutes)
  • Refractive index close to glass (1.510)
  • Chemically neutral, little sensitivity to water, no reaction to staining
  • Temperature limits: –17 to 42 °C
  • Not affected by ultraviolet light, non yellowing, stays optically clear for over 10 years
  • Spreads quickly and evenly without forming any air bubbles

Can be used in place of epoxy resins for mounting petrographic thin sections, used in fluorescence dyes in bones.

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Eukitt® mounting medium
  • Cat. No.:
    03989-500ML
  • M-Clarity quality level:
    MQ100

Specifications