2-Propanol, Supelco®
Glass bottle, 10 mL
Catalog # 91237-10ML-F
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0.786 g/cm³ (20 °C)
- Flash Pt:12 °C
- UN:1219
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:200-661-7
- REACH:01-2119457558-25
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:10 mL



