Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, EnvirElut™
Solid phase extraction cartridges, EnvirElut™
Straight barrel cartridge NH2, EnvirElut, 3 ml, 100/500 mg
Catalog # VARI12102158
Supplier:   VARIAN
Solid phase extraction cartridges, EnvirElut™
Catalog # VARI12102158
Supplier:  VARIAN
Solid phase extraction cartridges, EnvirElut™
Straight barrel cartridge NH2, EnvirElut, 3 ml, 100/500 mg
Price
383,62 €
Log in for availability
1 * 1 ST
Product Details & Documents
EnvirElut™ sorbents are specially designed for the extraction of a wide range of compounds from aqueous matrices. EnvirElut™ PAH and Pesticides are available in standard SPE straight barrel cartridges, which can be used on conventional vacuum manifolds such as the Vac Elut SPS 24.
  • Extreme purity offers cleanliness in extract
  • High capacity allows for the processing of large sample volumes
  • Broad compound specificity
Specifications
  • Capacity:
    3 ml
  • Description:
    Straight barrel cartridge NH2
  • Seq:
    01
  • Bed weight:
    100/500 mg
  • Phase:
    EnvirElut