Orders LinkContactUsLinkComponent
Graphite tubes and platforms
Graphite tubes and platforms
Solid pyrolytic graphite platforms, only for plateau tubes
Catalog # VARI6310001300
Supplier:   VARIAN
Graphite tubes and platforms
Catalog # VARI6310001300
Supplier:  VARIAN
Graphite tubes and platforms
Graphite tubes and platforms
Solid pyrolytic graphite platforms, only for plateau tubes
Price
462,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Graphite tubes are quality tested to ensure each batch passes our demanding performance specifications for contamination, sensitivity, precision, electrical resistance and lifetime.
  • High purity graphite reduces spurious absorption from trace contaminants and improves signal-to-noise
  • Every tube is hand-checked for reproducible and reliable results
  • Perform fewer repeats and increase productivity
Use partitioned tubes for general purpose work and when you need the best detection limits with samples that have low background. These are also ideal when measuring organic solutions, or samples with low viscosity that may spread throughout the tube, potentially degrading precision.
Omega platform tubes combine ease-of-use with perfect atomisation: the integrated platform ensures the perfect thermal profile to produce dense atom clouds for good signal-to-noise ratios and the best detection limits. The combination of the solid pyrolytic platforms and plateau tubes provide continuity with older methods, and gives the best platform effect. This ensures best performance with samples that have high background or interferences.
Specifications
  • Description:
    Solid pyrolytic graphite platforms, only for plateau tubes
  • Seq:
    03