Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solution pH 7, Phosphate buffer

Buffer solution pH 7, Phosphate buffer

Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution, 7,00, ±0,02, Phosphate, Plastic bottle, 500 mL
Catalog # ACRO258595000
Buffer solution pH 7, Phosphate buffer
Catalog # ACRO258595000
CAS Number:  
Buffer solution pH 7, Phosphate buffer

Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution, 7,00, ±0,02, Phosphate, Plastic bottle, 500 mL

CAS Number:  
Price
39,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phosphate
  • Pk:
    500 mL
  • Description:
    Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
  • pH accuracy:
    ±0,02
  • pH value:
    7,00
  • Cat. No.:
    ACRO258595000
  • Concentration:
    3.54 g potassium dihydrogen phosphate and 14.7 g i-sodium hydrogen phosphate per litre
  • Appearance:
    Clear colourless solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.