Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solution pH 4, citrate buffer

Buffer solution pH 4, citrate buffer

4,00, ±0,02, Citrate, Colourless, Plastic bottle, 500 mL
Catalog # ACRO258585000
Buffer solution pH 4, citrate buffer
Catalog # ACRO258585000
CAS Number:  
Buffer solution pH 4, citrate buffer

4,00, ±0,02, Citrate, Colourless, Plastic bottle, 500 mL

CAS Number:  
Price
39,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Citrate
  • Pk:
    500 mL
  • Colour:
    Colourless
  • pH accuracy:
    ±0,02
  • pH value:
    4,00

Specifications