Orders LinkContactUsLinkComponent
Blocking solution, BLOK™ Casein
Blocking solution, BLOK™ Casein
BLOK™ Casein in TBS, 1% solution, 2
Catalog # 786-196
Supplier:   G-Biosciences
Blocking solution, BLOK™ Casein
Catalog # 786-196
Supplier:  G-Biosciences
CAS Number:  
Blocking solution, BLOK™ Casein
Blocking solution, BLOK™ Casein
BLOK™ Casein in TBS, 1% solution, 2
CAS Number:  
Price
184,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
BLOK™ Casein is a purified protein, used for blocking free binding sites in Western blot, ELISA plate well, immunohistochemistry and other immunochemical applications.
  • Ready to use
  • Available in the choices of TBS or PBS buffers
Specifications
Specifications
  • Pk:
    2
  • Description:
    BLOK™ Casein in TBS, 1% solution
Specifications