Order Entry
Austria
Orders LinkContactUsLinkComponent
 

J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate

DILUT-IT™ concentrate, 9,00, ±0,02, Borate, Blue, 1
Catalog # 4862.
Supplier:   Avantor
J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate
Catalog # 4862.
Supplier:  Avantor
CAS Number:  
J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate

DILUT-IT™ concentrate, 9,00, ±0,02, Borate, Blue, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Volumetric concentrate, for dilution to 500 ml.

  • Traceable to NIST

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • pH buffer type:
    Borate
  • Pk:
    1
  • Colour:
    Blue
  • Description:
    DILUT-IT™ concentrate
  • pH accuracy:
    ±0,02
  • pH value:
    9,00

Specifications