Buffer PH 10,00 ( Blue ) 1 * 500 mL
Catalog # REGE1100525CTT
Supplier: REAGECON
Frequently Bought Together
Ammonium cerium(IV) sulphate dihydrate ≥94.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Viscosity standards
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Instrument trays
Supplier: BURKLE
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonia solution 17% Reag. Ph. Eur. 1004701
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Bulb pipettes, BLAUBRAND®, two marks, class AS
Supplier: Brand
REAGENT ACETIC ANHYDRIDE SOLUTION R1 1 * 1 L
Supplier: REAGECON
Buffer PH 4,00 1 * 500 mL
Supplier: REAGECON
BUFFER SOLUTION PH1.00+0.02 25GRAD 1 * 500 mL
Supplier: REAGECON