Tissue processing/embedding cassettes, Swingsette®, M517
Turquoise
Catalog # 720-1658
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M517
Catalog # 720-1658
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer.
- Hinged lid supplied in separate packaging to make it easier to use the cassettes with commercial printers; large tab to make opening easier
- Disposable plastic cassettes are highly resistant to histological solvents
- Oblong opening for good fluid exchange
- Front face inclined 45° so they can be used with certain automatic labelling systems
Verpackung: Each case contains two dispenser boxes of 500 cassettes each and one dispenser box of 1000 lids.
Specifications
- Colour:Turquoise
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



