Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Swingsette®, M515

Tissue processing/embedding cassettes, Swingsette®, M515

Green
Catalog # 720-1636
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M515
Catalog # 720-1636
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M515
Tissue processing/embedding cassettes, Swingsette®, M515

Green

Price
258,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

Acetal polymer.

  • Integral, hinged lid, with large tab for easy opening
  • Disposable plastic cassettes highly resistant to histological solvents
  • Flow-through slots maximise fluid exchange
  • Front face inclined 45° to enable use with leading automatic labelling systems.

Packaging: Each case contains three boxes of 500 cassettes.

Specifications
  • Colour:
    Green