Order Entry
Austria
Orders LinkContactUsLinkComponent
Flat canisters

Flat canisters

8 L, Natural, B63, With drain valve, autoclavable, 375×305×165 mm
Catalog # 590-0576
Supplier:   S.C.A.T.
Flat canisters
Catalog # 590-0576
Supplier:  S.C.A.T.
Flat canisters
Flat canisters

8 L, Natural, B63, With drain valve, autoclavable, 375×305×165 mm

Price
198,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Due to their flat design, SCAT flat canisters have been specially developed for use in confined spaces such as cupboards, shelves, drawers or refrigerators, as they are structurally low in height.

  • Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in laboratories
  • Available with B63 or S70/71 threads and can be adapted to many other thread sizes by means of an adapter
  • Available either as compact 8 L canister or as more voluminous 20 L canister

SCAT flat canisters are an ideal way of not having to dispense with safe storage even when space is limited, as they can be integrated into laboratory furniture to save space.

SCAT flat pack canisters are also optimally available with UN-Y approval for the storage of hazardous goods or with an integrated drain cock for safe emptying.

Specifications
  • Thread:
    B63
  • Type:
    With drain valve, autoclavable
  • Capacity:
    8 L
  • Colour:
    Natural
  • Seq:
    02
  • W×D×H:
    375×305×165 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.