ContactUsLinkComponent
Multi use logger with BioCote® antimicrobial protection, internal sensor, measurement range -30°C to +70°C, memory capacity up to 30 00 readings, critical data shown on display. 1 * 1 items
Catalog # COMAEVT2
Supplier:  COMARK
Multi use logger with BioCote® antimicrobial protection, internal sensor, measurement range -30°C to +70°C, memory capacity up to 30 00 readings, critical data shown on display. 1 * 1 items
Catalog # COMAEVT2
Supplier:  COMARK
Frequently Bought Together