Orders LinkContactUsLinkComponent
Cold traps, Type KF
Cold traps, Type KF
Cold trap S 29 GL-A, borosilicate glass 3,3, with threaded glass joint GL 18 and PTFE olive, 150 ml condensate capacity
Catalog # KGWA17377
Supplier:   KGW
Cold traps, Type KF
Catalog # KGWA17377
Supplier:  KGW
Cold traps, Type KF
Cold traps, Type KF
Cold trap S 29 GL-A, borosilicate glass 3,3, with threaded glass joint GL 18 and PTFE olive, 150 ml condensate capacity
Price
367,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminium
Specifications
  • Description:
    Cold trap S 29 GL-A, borosilicate glass 3,3, with threaded glass joint GL 18 and PTFE olive, 150 ml condensate capacity
  • Seq:
    05
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Plastic ring for cold trap, type KF
    Plastic ring for cold trap, type KF
    KGWA1733
    Log in for availability
    1 * 1 ST - 67,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Cooling finger S 29-GL
    Cooling finger S 29-GL
    KGWA1741
    Log in for availability
    1 * 1 ST - 298,00 €