Orders LinkContactUsLinkComponent
VWR®, Objectives for Microscope
VWR®, Objectives for Microscope
Semi plan achromatic objective
Catalog # 630-0843
Supplier:   VWR Collection
VWR®, Objectives for Microscope
Catalog # 630-0843
Supplier:  VWR Collection
VWR®, Objectives for Microscope
Semi plan achromatic objective
Price
63,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    20×
  • Description:
    Semi plan achromatic objective
  • Seq:
    03